siRNA Details
Gene Symbol: TTYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201461.1 | NP_001188390.1 | 12 | 1420 | ||
NM_020659.3 | NP_065710.1 | 12 | 1420 | |||
XM_017027026.1 | XP_016882515.1 | 12 | 1264 | |||
GenBank mRNA | AB188496.1 | - | 12 | 1420 | ||
AF177909.1 | - | 12 | 1334 | |||
AK293468.1 | - | 13 | 1451 | |||
BC019358.1 | - | 9 | 753 | 3960435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
