siRNA Details
Gene Symbol: TTYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020659.3 | NP_065710.1 | 12, 13 | 1429 | ||
XM_017027026.1 | XP_016882515.1 | 12, 13 | 1273 | |||
GenBank mRNA | AF177909.1 | - | 12, 13 | 1343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
