siRNA Details
Gene Symbol: TAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286074.1 | NP_001273003.1 | 4 | 594 | ||
NM_004606.4 | NP_004597.2 | 4 | 594 | |||
NM_138923.3 | NP_620278.1 | 4 | 594 | |||
NR_104387.1 | 4 | 594 | ||||
NR_104388.1 | 4 | 594 | ||||
NR_104389.1 | 4 | 594 | ||||
NR_104390.1 | 4 | 594 | ||||
NR_104391.1 | 4 | 594 | ||||
NR_104392.1 | 4 | 594 | ||||
NR_104393.1 | 4 | 594 | ||||
NR_104394.1 | 4 | 594 | ||||
NR_104395.1 | 4 | 594 | ||||
XM_005262295.1 | XP_005262352.1 | 4 | 528 | |||
XM_005262296.1 | XP_005262353.1 | 4 | 528 | |||
XM_005262297.4 | XP_005262354.1 | 4 | 613 | |||
XM_005262300.2 | XP_005262357.1 | 4 | 528 | |||
XM_006724682.3 | XP_006724745.1 | 3 | 1472 | |||
GenBank mRNA | AB300418.1 | - | 4 | 521 | ||
AK310201.1 | - | 4 | 530 | |||
AW937788.1 | - | 3 | 337 | |||
D90359.1 | - | 4 | 569 | |||
X07024.1 | - | 3 | 681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
