siRNA Details
Gene Symbol: NLRC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199138.1 | NP_001186067.1 | 4 | 664 | ||
NM_001199139.1 | NP_001186068.1 | 4 | 547 | |||
NM_021209.4 | NP_067032.3 | 4 | 678 | |||
XM_017004619.1 | XP_016860108.1 | 4 | 664 | |||
GenBank mRNA | AK095467.1 | - | 4 | 664 | ||
AK292673.1 | - | 4 | 649 | |||
AK314762.1 | - | 4 | 649 | |||
AY027789.1 | - | 4 | 679 | |||
AY032589.1 | - | 3 | 438 | |||
AY035391.1 | - | 4 | 547 | |||
AY358152.1 | - | 4 | 626 | |||
BC031555.1 | - | 4 | 634 | 5179909 |
