siRNA Details
Gene Symbol: NTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021229.3 | NP_067052.2 | 10 | 2249 | ||
GenBank mRNA | AF119916.3 | - | 11 | 1786 | ||
AF278532.1 | - | 10 | 2251 | |||
AF297711.1 | - | 10 | 2101 | |||
AK024691.1 | - | 9 | 1325 | |||
AK026833.1 | - | 5 | 595 | |||
AK226070.1 | - | 10 | 2106 | |||
AK290591.1 | - | 10 | 1786 | |||
AK315796.1 | - | 10 | 2108 | |||
AL137540.1 | - | 8 | 1117 | |||
AL833767.1 | - | 9 | 1653 | |||
AY330211.1 | - | 9 | 1731 | |||
BC013591.1 | - | 9 | 1324 | |||
BC136798.1 | - | 10 | 2224 | 9020788 | ||
BC136799.1 | - | 10 | 2224 | 9020789 |
