siRNA Details
Gene Symbol: HMGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003483.4 | NP_003474.1 | 4, 5 | 1104 | ||
GenBank mRNA | AF533653.1 | - | 196 | |||
AI290477.1 | - | 4, 5 | 330 | |||
U28749.1 | - | 4, 5 | 1104 | |||
X92518.1 | - | 4, 5 | 1603 | |||
Z31595.1 | - | 4, 5 | 1105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
