siRNA Details
Gene Symbol: PRDM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024504.3 | NP_078780.1 | 4 | 1063 | ||
XM_011517572.1 | XP_011515874.1 | 3 | 343 | |||
GenBank mRNA | AF319458.1 | - | 4 | 1063 | ||
AK022595.1 | - | 4 | 1063 | |||
BC052311.1 | - | 4 | 976 | 6671984 | ||
KJ463837.1 | - | 3 | 861 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
