siRNA Details
Gene Symbol: DENND1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024820.2 | NP_079096.2 | 21 | 1838 | ||
XM_005252113.4 | XP_005252170.1 | 21 | 2643 | |||
XM_011518887.2 | XP_011517189.1 | 22 | 2700 | |||
GenBank mRNA | AK024782.1 | - | 21 | 1826 | ||
AK225104.1 | - | 21 | 1826 | |||
BC009616.2 | - | 20 | 1806 | |||
BC039703.1 | - | 21 | 1691 | 4999484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
