siRNA Details
Gene Symbol: LINC00158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024027.2 | 5 | 1078 | |||
GenBank mRNA | AY035382.1 | - | 4 | 981 | ||
AY035383.1 | - | 5 | 1077 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024027.2 | 5 | 1078 | |||
GenBank mRNA | AY035382.1 | - | 4 | 981 | ||
AY035383.1 | - | 5 | 1077 |