siRNA Details
Gene Symbol: ZNF398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020781.3 | NP_065832.1 | 7 | 4075 | ||
NM_170686.2 | NP_733787.1 | 6 | 4012 | |||
XM_011516439.2 | XP_011514741.1 | 7 | 3893 | |||
XM_011516440.2 | XP_011514742.1 | 6 | 3865 | |||
XM_011516441.2 | XP_011514743.1 | 6 | 3848 | |||
GenBank mRNA | AB037760.1 | - | 2 | 3033 | ||
AY049743.1 | - | 7 | 4075 | |||
AY049744.1 | - | 6 | 3754 |
