siRNA Details
Gene Symbol: LUC7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320226.1 | NP_001307155.1 | 2, 3 | 306 | ||
NM_018032.4 | NP_060502.1 | 2, 3 | 306 | |||
NM_201412.2 | NP_958815.1 | 2, 3 | 306 | |||
XM_005255427.3 | XP_005255484.1 | 3, 4 | 343 | |||
XM_005255429.3 | XP_005255486.1 | 3, 4 | 325 | |||
XM_017023437.1 | XP_016878926.1 | 2, 3 | 1369 | |||
XM_017023438.1 | XP_016878927.1 | 2, 3 | 1369 | |||
XM_017023439.1 | XP_016878928.1 | 2, 3 | 1369 | |||
XM_017023440.1 | XP_016878929.1 | 2, 3 | 1369 | |||
GenBank mRNA | AJ404326.1 | - | 2, 3 | 234 | ||
AK001093.1 | - | 2, 3 | 234 | |||
AK223084.1 | - | 2, 3 | 237 | |||
AY005111.1 | - | 2, 3 | 274 | |||
BC003194.1 | - | 2, 3 | 251 | 3536109 | ||
BC065198.1 | - | 2, 3 | 1523 | 5212471 | ||
BX647161.1 | - | 2, 3 | 1304 | |||
Z84716.2 | - | 1, 2 | 657 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028190, NM_025881, BC034135, AK160293, AK007781, and others.
Gene Ontology Categories:
