siRNA Details
Gene Symbol: COPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290033.1 | NP_001276962.1 | 19 | 2045 | ||
NM_012133.5 | NP_036265.3 | 19 | 2045 | |||
GenBank mRNA | AB047847.1 | - | 19 | 1955 | ||
AF157833.1 | - | 19 | 2015 | |||
AF207598.1 | - | 19 | 2011 | |||
AK002086.1 | - | 11 | 1347 | |||
AK022934.1 | - | 19 | 1942 | |||
AK024414.1 | - | 12 | 1423 | |||
AK291409.1 | - | 19 | 2035 | |||
AL162012.1 | - | 2 | 177 | |||
BC017443.1 | - | 19 | 1938 | |||
BC110796.1 | - | 19 | 1992 | 5197024 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
