siRNA Details
Gene Symbol: FBXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171713.1 | NP_001165184.1 | 6, 7 | 435 | ||
NM_012157.3 | NP_036289.3 | 6, 7 | 438 | |||
XM_005265014.2 | XP_005265071.1 | 6, 7 | 458 | |||
XM_005265015.3 | XP_005265072.1 | 7 | 678 | |||
XM_005265016.2 | XP_005265073.1 | 7 | 422 | |||
XM_005265017.3 | XP_005265074.1 | 7 | 454 | |||
XM_011533559.2 | XP_011531861.1 | 8 | 623 | |||
XM_017006092.1 | XP_016861581.1 | 8 | 515 | |||
XM_017006093.1 | XP_016861582.1 | 7 | 505 | |||
XM_017006094.1 | XP_016861583.1 | 8 | 538 | |||
GenBank mRNA | AF174589.1 | - | 7, 8 | 409 | ||
AF176518.1 | - | 6, 7 | 373 | |||
AF186273.1 | - | 6, 7 | 388 | |||
AK001271.1 | - | 6, 7 | 438 | |||
AK001438.1 | - | 6, 7 | 406 | |||
AK294209.1 | - | 3, 4 | 189 | |||
AK297534.1 | - | 7 | 422 | |||
AK298967.1 | - | 6, 7 | 404 | |||
AK299665.1 | - | 7, 8 | 521 | |||
AK302218.1 | - | 8, 9 | 666 | |||
AK303764.1 | - | 4, 5 | 298 | |||
AK316229.1 | - | 6, 7 | 459 | |||
AK316383.1 | - | 7, 8 | 578 | |||
AU124816.1 | - | 6, 7 | 438 | |||
BC031556.1 | - | 6, 7 | 382 | 5197151 | ||
CR457121.1 | - | 6, 7 | 367 |
