siRNA Details
Gene Symbol: OGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181672.2 | NP_858058.1 | 6 | 957 | ||
NM_181673.2 | NP_858059.1 | 6 | 927 | |||
GenBank mRNA | AF223393.1 | - | 2 | 594 | ||
AK313530.1 | - | 2 | 873 | |||
AL833085.2 | - | 6 | 937 | |||
BC014434.1 | - | 6 | 858 | 4865031 | ||
BC038180.1 | - | 6 | 890 | 5017795 | ||
BX537844.1 | - | 6 | 915 | |||
U77413.1 | - | 2 | 606 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
