siRNA Details
Gene Symbol: LSM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130710.1 | NP_001124182.1 | 5 | 660 | ||
NM_001139499.1 | NP_001132971.1 | 5 | 899 | |||
NM_012322.2 | NP_036454.1 | 5 | 723 | |||
NR_024466.1 | 5 | 1470 | ||||
GenBank mRNA | AF182291.1 | - | 5 | 684 | ||
AJ238097.1 | - | 5 | 678 | |||
AK024217.1 | - | 5 | 706 |
