siRNA Details
Gene Symbol: RBMS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177711.1 | NP_001171182.1 | 11, 12 | 1423 | ||
NM_001177712.1 | NP_001171183.1 | 12, 13 | 1462 | |||
NM_014483.3 | NP_055298.2 | 11, 12 | 1423 | |||
XM_017006179.1 | XP_016861668.1 | 11, 12 | 1872 | |||
XM_017006180.1 | XP_016861669.1 | 10, 11 | 1824 | |||
XM_017006181.1 | XP_016861670.1 | 10, 11 | 1821 | |||
XM_017006182.1 | XP_016861671.1 | 11, 12 | 1900 | |||
GenBank mRNA | AF023259.1 | - | 11, 12 | 1200 | ||
AK292789.1 | - | 11, 12 | 1099 | |||
BC117315.1 | - | 12, 13 | 1159 | 40125866 | ||
BC143521.1 | - | 12, 13 | 1159 | 9052030 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
