siRNA Details
Gene Symbol: ZNF330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292002.1 | NP_001278931.1 | 8, 9 | 856 | ||
NM_014487.5 | NP_055302.1 | 9, 10 | 927 | |||
XM_017008033.1 | XP_016863522.1 | 9, 10 | 1071 | |||
GenBank mRNA | AJ006591.2 | - | 9, 10 | 872 | ||
AK002106.1 | - | 9, 10 | 876 | |||
AK223609.1 | - | 9, 10 | 878 | |||
AK294089.1 | - | 8, 9 | 810 | |||
AK315463.1 | - | 9, 10 | 913 | |||
BC004421.1 | - | 9, 10 | 903 | 3352916 |
