siRNA Details
Gene Symbol: ZDHHC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015457.2 | NP_056272.2 | 3 | 1376 | ||
XM_011544899.1 | XP_011543201.1 | 3 | 1306 | |||
XM_011544900.1 | XP_011543202.1 | 3 | 1214 | |||
XM_011544901.1 | XP_011543203.1 | 3 | 1337 | |||
GenBank mRNA | AB051535.1 | - | 3 | 1368 | ||
AK023130.1 | - | 2 | 533 | |||
AY894889.1 | - | 3 | 1482 | |||
BC026967.1 | - | 2 | 119 | 3913317 | ||
BX647707.1 | - | 2 | 237 |
