siRNA Details
Gene Symbol: ZNF385A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130967.2 | NP_001124439.1 | 5 | 582 | ||
NM_001290002.1 | NP_001276931.1 | 6 | 772 | |||
NM_001290004.1 | NP_001276933.1 | 6 | 727 | |||
NM_015481.2 | NP_056296.1 | 4 | 677 | |||
XM_005268783.4 | XP_005268840.1 | 5 | 604 | |||
XM_006719342.2 | XP_006719405.1 | 6 | 775 | |||
XM_011538168.2 | XP_011536470.1 | 6 | 694 | |||
XM_011538169.2 | XP_011536471.1 | 5 | 598 | |||
XM_011538170.1 | XP_011536472.1 | 4 | 447 | |||
XM_017019174.1 | XP_016874663.1 | 5 | 910 | |||
GenBank mRNA | AB593085.1 | - | 6 | 637 | ||
AF304052.1 | - | 4 | 644 | |||
AK092724.1 | - | 5 | 451 | |||
AK315613.1 | - | 5 | 575 | |||
AL117462.1 | - | 3 | 287 | |||
AY461717.1 | - | 6 | 752 | |||
BC029752.1 | - | 5 | 563 | 4476647 | ||
BM546460.1 | - | 602 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013866, AF118566, CK619761, AK137748, BC017644.
Gene Ontology Categories:
