siRNA Details
Gene Symbol: ZNF385A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130967.2 | NP_001124439.1 | 5, 6 | 720 | ||
NM_001290002.1 | NP_001276931.1 | 6, 7 | 910 | |||
NM_001290004.1 | NP_001276933.1 | 6, 7 | 865 | |||
NM_015481.2 | NP_056296.1 | 4, 5 | 815 | |||
XM_005268783.4 | XP_005268840.1 | 5, 6 | 742 | |||
XM_006719342.2 | XP_006719405.1 | 6, 7 | 913 | |||
XM_011538168.2 | XP_011536470.1 | 6, 7 | 832 | |||
XM_011538169.2 | XP_011536471.1 | 5, 6 | 736 | |||
XM_011538170.1 | XP_011536472.1 | 4, 5 | 585 | |||
XM_017019174.1 | XP_016874663.1 | 5, 6 | 1048 | |||
GenBank mRNA | AB593085.1 | - | 6, 7 | 775 | ||
AF304052.1 | - | 4, 5 | 782 | |||
AK092724.1 | - | 5, 6 | 589 | |||
AK315613.1 | - | 5, 6 | 713 | |||
AL117462.1 | - | 3, 4 | 425 | |||
AY461717.1 | - | 6, 7 | 890 | |||
BC029752.1 | - | 5, 6 | 701 | 4476647 | ||
BM546460.1 | - | 740 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013866, AF118566, AK137748, BC017644.
Gene Ontology Categories:
