siRNA Details
Gene Symbol: RWDD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199682.1 | NP_001186611.1 | 3, 4 | 726 | ||
NM_001278247.1 | NP_001265176.1 | 4, 5 | 859 | |||
NM_001278248.1 | NP_001265177.1 | 4, 5 | 906 | |||
NM_015485.4 | NP_056300.2 | 3, 4 | 773 | |||
NR_103483.1 | 3, 4 | 790 | ||||
NR_103484.1 | 4, 5 | 406 | ||||
GenBank mRNA | BC010936.2 | - | 3, 4 | 726 | ||
BC125142.1 | - | 4, 5 | 807 | 40119943 | ||
BU858771.1 | - | 2, 3 | 280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
