siRNA Details
Gene Symbol: NBEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015678.4 | NP_056493.3 | 6 | 1404 | ||
XM_005266346.4 | XP_005266403.1 | 6 | 1404 | |||
XM_005266347.4 | XP_005266404.2 | 6 | 1404 | |||
XM_005266348.4 | XP_005266405.2 | 6 | 1404 | |||
XM_006719803.3 | XP_006719866.1 | 6 | 1404 | |||
XM_006719805.3 | XP_006719868.1 | 6 | 1404 | |||
XM_006719806.3 | XP_006719869.1 | 6 | 1404 | |||
XM_011535047.2 | XP_011533349.1 | 6 | 1404 | |||
XM_017020544.1 | XP_016876033.1 | 6 | 1404 | |||
XM_017020545.1 | XP_016876034.1 | 6 | 1404 | |||
XM_017020546.1 | XP_016876035.1 | 6 | 692 | |||
GenBank mRNA | AF467288.1 | - | 7 | 1076 |
