siRNA Details
Gene Symbol: ATP5S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015684.3 | NP_056499.2 | 3 | 1134 | ||
GenBank mRNA | AF052186.1 | - | 3 | 1086 | ||
U79253.1 | - | 3 | 1085 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
