siRNA Details
Gene Symbol: ZNF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006956.2 | NP_008887.2 | 6 | 2718 | ||
NM_016265.3 | NP_057349.2 | 5 | 2832 | |||
GenBank mRNA | AB021643.1 | - | 6 | 2244 | ||
AF505656.1 | - | 5 | 2497 | |||
AK022691.1 | - | 1 | 1682 | |||
BC051892.1 | - | 6 | 2706 | |||
BC154414.1 | - | 5 | 2820 | 40148925 | ||
BX640749.1 | - | 5 | 2243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
