siRNA Details
Gene Symbol: IP6K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005909.2 | NP_001005909.1 | 3 | 496 | ||
NM_016291.3 | NP_057375.2 | 3 | 499 | |||
XM_006713199.3 | XP_006713262.1 | 4 | 603 | |||
XM_006713200.1 | XP_006713263.1 | 4 | 573 | |||
XM_006713201.1 | XP_006713264.1 | 4 | 650 | |||
XM_006713202.1 | XP_006713265.1 | 4 | 652 | |||
XM_011533816.1 | XP_011532118.1 | 4 | 633 | |||
XM_011533817.1 | XP_011532119.1 | 5 | 1538 | |||
XM_011533818.1 | XP_011532120.1 | 4 | 1806 | |||
XM_017006583.1 | XP_016862072.1 | 3 | 495 | |||
XM_017006584.1 | XP_016862073.1 | 3 | 470 | |||
XM_017006585.1 | XP_016862074.1 | 4 | 585 | |||
XM_017006586.1 | XP_016862075.1 | 4 | 2446 | |||
XM_017006587.1 | XP_016862076.1 | 7 | 1795 | |||
GenBank mRNA | AF177145.1 | - | 2 | 393 | ||
AK024045.1 | - | 3 | 436 | |||
AK315204.1 | - | 3 | 437 | |||
AL117458.1 | - | 1 | 23 | |||
AY007091.1 | - | 3 | 422 | |||
BC019694.1 | - | 3 | 408 | 4941646 | ||
BC065533.1 | - | 3 | 425 | 6162048 | ||
DB039955.1 | - | 3 | 496 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
