siRNA Details
Gene Symbol: IP6K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005909.2 | NP_001005909.1 | 4 | 773 | ||
NM_016291.3 | NP_057375.2 | 4 | 776 | |||
XM_006713199.3 | XP_006713262.1 | 5 | 880 | |||
XM_006713200.1 | XP_006713263.1 | 5 | 850 | |||
XM_006713201.1 | XP_006713264.1 | 5 | 927 | |||
XM_006713202.1 | XP_006713265.1 | 5 | 929 | |||
XM_011533816.1 | XP_011532118.1 | 5 | 910 | |||
XM_011533817.1 | XP_011532119.1 | 6 | 1815 | |||
XM_011533818.1 | XP_011532120.1 | 5 | 2083 | |||
XM_017006583.1 | XP_016862072.1 | 4 | 772 | |||
XM_017006584.1 | XP_016862073.1 | 4 | 747 | |||
XM_017006585.1 | XP_016862074.1 | 5 | 862 | |||
XM_017006586.1 | XP_016862075.1 | 5 | 2723 | |||
XM_017006587.1 | XP_016862076.1 | 8 | 2072 | |||
GenBank mRNA | AF177145.1 | - | 3 | 670 | ||
AK024045.1 | - | 4 | 713 | |||
AK315204.1 | - | 4 | 714 | |||
AL117458.1 | - | 2 | 300 | |||
AY007091.1 | - | 4 | 699 | |||
BC019694.1 | - | 4 | 685 | 4941646 | ||
BC065533.1 | - | 4 | 702 | 6162048 |

More Information
Other Species Targeted

Gene Ontology Categories:
