siRNA Details
Gene Symbol: COPS7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164093.1 | NP_001157565.1 | 3 | 690 | ||
NM_001164094.1 | NP_001157566.1 | 4 | 555 | |||
NM_001164095.2 | NP_001157567.1 | 4 | 626 | |||
NM_016319.3 | NP_057403.1 | 4 | 528 | |||
XM_005253694.2 | XP_005253751.1 | 4 | 453 | |||
GenBank mRNA | AB014764.1 | - | 3 | 316 | ||
AB033603.1 | - | 4 | 393 | |||
AF193844.1 | - | 3 | 316 | |||
AF210052.1 | - | 4 | 457 | |||
AK001318.1 | - | 4 | 440 | |||
AK021721.1 | - | 4 | 493 | |||
AK022488.1 | - | 4 | 435 | |||
AK292621.1 | - | 3 | 686 | |||
BC011789.2 | - | 4 | 416 | 4025790 | ||
BC093015.1 | - | 4 | 467 | 30344094 | ||
DA365275.1 | - | 4 | 500 |
