siRNA Details
Gene Symbol: PSMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF417108.1 | - | 2 | 183 | ||
AJ006291.1 | - | 3 | 353 | |||
AK308614.1 | - | 2 | 290 | |||
AY463963.1 | - | 3 | 290 | |||
BE779375.1 | - | 3 | 417 | |||
CB160221.1 | - | 3 | 281 | |||
CR541821.1 | - | 3 | 290 | |||
CR541852.1 | - | 3 | 290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
