siRNA Details
Gene Symbol: TSNAX-DISC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028393.1 | 11 | 2411 | |||
NR_028394.1 | 13 | 2594 | ||||
NR_028396.1 | 12 | 2466 | ||||
NR_028397.1 | 12 | 2331 | ||||
NR_028398.1 | 10 | 2085 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
