siRNA Details
Gene Symbol: DAZAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018959.3 | NP_061832.2 | 3, 4 | 434 | ||
NM_170711.2 | NP_733829.1 | 3, 4 | 434 | |||
XM_005259531.3 | XP_005259588.1 | 3, 4 | 387 | |||
XM_005259534.3 | XP_005259591.1 | 3, 4 | 393 | |||
XM_005259535.3 | XP_005259592.1 | 3, 4 | 395 | |||
XM_005259536.3 | XP_005259593.1 | 3, 4 | 395 | |||
XM_011527904.2 | XP_011526206.1 | 3, 4 | 1699 | |||
XM_011527905.2 | XP_011526207.1 | 3, 4 | 1707 | |||
XM_011527906.2 | XP_011526208.1 | 3, 4 | 1707 | |||
XM_011527907.2 | XP_011526209.1 | 3, 4 | 1716 | |||
XM_011527908.2 | XP_011526210.1 | 3, 4 | 1706 | |||
XM_011527909.2 | XP_011526211.1 | 3, 4 | 1706 | |||
XM_011527910.2 | XP_011526212.1 | 3, 4 | 786 | |||
XM_017026581.1 | XP_016882070.1 | 3, 4 | 429 | |||
XM_017026582.1 | XP_016882071.1 | 3, 4 | 1722 | |||
GenBank mRNA | AF181719.1 | - | 3, 4 | 337 | ||
AK056850.1 | - | 1, 2 | 57 | |||
AK125032.1 | - | 2, 3 | 1730 | |||
AL360261.1 | - | 3, 4 | 329 | |||
BC012062.1 | - | 3, 4 | 302 | 4549444 | ||
BI909277.1 | - | 3, 4 | 418 | |||
BU557183.1 | - | 434 | ||||
KU178593.1 | - | 3, 4 | 229 |
