siRNA Details
Gene Symbol: OASL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003733.3 | NP_003724.1 | 4 | 1054 | ||
XM_017020140.1 | XP_016875629.1 | 4 | 789 | |||
GenBank mRNA | AJ225089.1 | - | 4 | 789 | ||
AK023956.1 | - | 4 | 790 | |||
AK314419.1 | - | 4 | 789 | |||
BC117408.1 | - | 4 | 807 | 40125959 | ||
BC117410.1 | - | 4 | 807 | 40125961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
