siRNA Details
Gene Symbol: SOST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF170491.1 | - | 1 | 1121 | ||
AF326739.1 | - | 2 | 1956 | |||
AY358627.1 | - | 2 | 1970 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
