siRNA Details
Gene Symbol: BTBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025238.3 | NP_079514.1 | 5, 6 | 1251 | ||
GenBank mRNA | AF257241.1 | - | 5, 6 | 1130 | ||
AF355402.1 | - | 5, 6 | 1131 | |||
AF529369.1 | - | 5, 6 | 1048 | |||
AK000731.1 | - | 5, 6 | 1115 | |||
AK225355.1 | - | 5, 6 | 1115 | |||
AK225362.1 | - | 5, 6 | 1115 | |||
AK298534.1 | - | 2, 3 | 398 | |||
AL136853.1 | - | 5, 6 | 1131 | |||
AY364253.1 | - | 5, 6 | 1048 | |||
BC017464.1 | - | 5, 6 | 1078 | |||
BC028097.1 | - | 5, 6 | 1151 | 5242418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
