siRNA Details
Gene Symbol: MLXIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF245470.1 | - | 17 | 2945 | ||
AF245471.1 | - | 17 | 2888 | |||
AF245472.1 | - | 17 | 2939 | |||
AF245473.1 | - | 17 | 2882 | |||
AF245474.1 | - | 16 | 2794 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
