siRNA Details
Gene Symbol: MRPS31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005830.3 | NP_005821.2 | 4 | 774 | ||
GenBank mRNA | AK315248.1 | - | 4 | 732 | ||
BC022045.1 | - | 4 | 742 | 4774270 | ||
Z68747.1 | - | 4 | 704 |
Gene Symbol: HNRNPA1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011724.1 | NP_001011724.1 | 4 | 730 | ||
GenBank mRNA | AK094423.1 | - | 3 | 480 | ||
AK123081.1 | - | 4 | 1123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
