siRNA Details
Gene Symbol: TRIM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006470.3 | NP_006461.3 | 7 | 1449 | ||
GenBank mRNA | AF096870.1 | - | 4 | 1119 | ||
AK002035.1 | - | 4 | 915 | |||
AK056026.1 | - | 5 | 1730 | |||
AK314557.1 | - | 4 | 929 | |||
BC001564.1 | - | 4 | 779 | |||
BC015674.2 | - | 3 | 468 | |||
BC053514.1 | - | 7 | 1449 | 6263729 | ||
BC067096.1 | - | 7 | 1449 | 6088764 | ||
CR457136.1 | - | 4 | 892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
