siRNA Details
Gene Symbol: ZMYND11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202464.1 | NP_001189393.1 | 14 | 2574 | ||
NM_001202465.1 | NP_001189394.1 | 13 | 2814 | |||
NM_001202466.1 | NP_001189395.1 | 14 | 2904 | |||
NM_006624.5 | NP_006615.2 | 15 | 2982 | |||
XM_005252359.4 | XP_005252416.1 | 15 | 2822 | |||
XM_005252361.3 | XP_005252418.1 | 14 | 2743 | |||
XM_005252362.2 | XP_005252419.1 | 14 | 2907 | |||
XM_006717376.2 | XP_006717439.1 | 14 | 2945 | |||
XM_017015587.1 | XP_016871076.1 | 15 | 2735 | |||
XM_017015588.1 | XP_016871077.1 | 15 | 2971 | |||
XM_017015589.1 | XP_016871078.1 | 15 | 2812 | |||
XM_017015590.1 | XP_016871079.1 | 15 | 3269 | |||
XM_017015591.1 | XP_016871080.1 | 16 | 2782 | |||
XM_017015592.1 | XP_016871081.1 | 14 | 2650 | |||
XM_017015593.1 | XP_016871082.1 | 14 | 3090 | |||
XM_017015594.1 | XP_016871083.1 | 14 | 2578 | |||
GenBank mRNA | BC091489.1 | - | 14 | 2574 | 6186814 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
