siRNA Details
Gene Symbol: ZMYND11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202464.1 | NP_001189393.1 | 14 | 2681 | ||
NM_001202465.1 | NP_001189394.1 | 13 | 2921 | |||
NM_001202466.1 | NP_001189395.1 | 14 | 3011 | |||
NM_006624.5 | NP_006615.2 | 15 | 3089 | |||
XM_005252359.4 | XP_005252416.1 | 15 | 2929 | |||
XM_005252361.3 | XP_005252418.1 | 14 | 2850 | |||
XM_005252362.2 | XP_005252419.1 | 14 | 3014 | |||
XM_006717376.2 | XP_006717439.1 | 14 | 3052 | |||
XM_017015587.1 | XP_016871076.1 | 15 | 2842 | |||
XM_017015588.1 | XP_016871077.1 | 15 | 3078 | |||
XM_017015589.1 | XP_016871078.1 | 15 | 2919 | |||
XM_017015590.1 | XP_016871079.1 | 15 | 3376 | |||
XM_017015591.1 | XP_016871080.1 | 16 | 2889 | |||
XM_017015592.1 | XP_016871081.1 | 14 | 2757 | |||
XM_017015593.1 | XP_016871082.1 | 14 | 3197 | |||
XM_017015594.1 | XP_016871083.1 | 14 | 2685 | |||
GenBank mRNA | BC091489.1 | - | 14 | 2681 | 6186814 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
