siRNA Details
Gene Symbol: UTP14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166221.1 | NP_001159693.1 | 3 | 243 | ||
NM_006649.3 | NP_006640.2 | 3 | 243 | |||
XM_017029237.1 | XP_016884726.1 | 3 | 177 | |||
XM_017029238.1 | XP_016884727.1 | 3 | 235 | |||
GenBank mRNA | AK291918.1 | - | 3 | 210 | ||
AK298578.1 | - | 3 | 212 | |||
AK304563.1 | - | 3 | 173 | |||
BC001149.1 | - | 3 | 179 | 3355662 | ||
BC009649.1 | - | 3 | 181 | 3459192 | ||
BC014987.1 | - | 3 | 171 | 4901477 | ||
BP353997.1 | - | 3 | 241 | |||
BU942896.1 | - | 3 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
