siRNA Details
Gene Symbol: UTP14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021645.5 | NP_067677.4 | 2 | 929 | ||
GenBank mRNA | BC089407.1 | - | 1 | 338 | 3065365 | |
BC122536.1 | - | 2 | 706 | 40036054 | ||
D87455.1 | - | 2 | 929 |
Gene Symbol: ALG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004127.2 | NP_001004127.2 | 4 | 1921 | ||
NR_036571.2 | 2 | 758 | ||||
GenBank mRNA | AK025456.1 | - | 4 | 1894 | ||
AK296747.1 | - | 4 | 1911 | |||
BC073862.1 | - | 2 | 1077 |
Gene Symbol: UTP14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166221.1 | NP_001159693.1 | 4 | 289 | ||
NM_006649.3 | NP_006640.2 | 4 | 289 | |||
XM_017029237.1 | XP_016884726.1 | 4 | 223 | |||
XM_017029238.1 | XP_016884727.1 | 4 | 281 | |||
GenBank mRNA | AK291918.1 | - | 4 | 256 | ||
AK298578.1 | - | 4 | 258 | |||
BC001149.1 | - | 4 | 225 | 3355662 | ||
BC009649.1 | - | 4 | 227 | 3459192 | ||
BC014987.1 | - | 4 | 217 | 4901477 | ||
BP353997.1 | - | 4 | 287 | |||
BU942896.1 | - | 4 | 291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
