siRNA Details
Gene Symbol: SMR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006685.3 | NP_006676.1 | 3 | 353 | ||
GenBank mRNA | BC015327.1 | - | 3 | 288 | 4397938 | |
BC144529.1 | - | 2 | 231 | 9053056 | ||
BX487896.1 | - | 3 | 303 | |||
CB958582.1 | - | 3 | 356 | |||
D29833.2 | - | 3 | 290 |
Gene Symbol: SMR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CD722466.1 | - | 4 | 345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
