siRNA Details
Gene Symbol: ASCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284271.1 | NP_001271200.1 | 11 | 2222 | ||
NM_006828.3 | NP_006819.2 | 11 | 2222 | |||
XM_011535394.2 | XP_011533696.1 | 11 | 2232 | |||
XM_011535395.2 | XP_011533697.1 | 12 | 2457 | |||
XM_011535396.2 | XP_011533698.1 | 10 | 2066 | |||
XM_017010205.1 | XP_016865694.1 | 12 | 2478 | |||
XM_017010206.1 | XP_016865695.1 | 12 | 2256 | |||
GenBank mRNA | AK299117.1 | - | 10 | 2071 | ||
AL834463.1 | - | 10 | 1907 | |||
AY013288.1 | - | 7 | 1041 | |||
BC026066.1 | - | 11 | 2219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
