siRNA Details
Gene Symbol: ASCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006828.3 | NP_006819.2 | 16 | 2924 | ||
XM_011535394.2 | XP_011533696.1 | 16 | 2934 | |||
XM_011535395.2 | XP_011533697.1 | 17 | 3159 | |||
XM_011535396.2 | XP_011533698.1 | 15 | 2768 | |||
XM_017010205.1 | XP_016865694.1 | 17 | 3180 | |||
XM_017010206.1 | XP_016865695.1 | 17 | 2958 | |||
GenBank mRNA | AK299117.1 | - | 15 | 2773 | ||
AL834463.1 | - | 15 | 2609 | |||
AY013288.1 | - | 12 | 1743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
