siRNA Details
Gene Symbol: RBPMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006867.3 | NP_006858.1 | 8 | 1264 | ||
XM_017012980.1 | XP_016868469.1 | 9 | 1396 | |||
XM_017012983.1 | XP_016868472.1 | 9 | 2382 | |||
XM_017012985.1 | XP_016868474.1 | 6 | 825 | |||
XM_017012990.1 | XP_016868479.1 | 7 | 1128 | |||
XM_017012991.1 | XP_016868480.1 | 10 | 1543 | |||
GenBank mRNA | D84110.1 | - | 8 | 1238 | ||
N88549.1 | - | 1 | 19 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
