siRNA Details
Gene Symbol: ZPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159878.1 | NP_001153350.1 | 8 | 1121 | ||
NM_007009.2 | NP_008940.2 | 8 | 1124 | |||
GenBank mRNA | BC005223.1 | - | 8 | 1083 | 3950015 | |
D17570.1 | - | 8 | 1090 |
