siRNA Details
Gene Symbol: CNTRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007018.4 | NP_008949.4 | 7 | 1137 | ||
XM_005251679.3 | XP_005251736.1 | 9 | 1409 | |||
XM_006716940.2 | XP_006717003.1 | 9 | 1409 | |||
XM_006716944.3 | XP_006717007.1 | 9 | 1409 | |||
XM_011518166.1 | XP_011516468.1 | 9 | 1409 | |||
XM_011518167.1 | XP_011516469.1 | 8 | 1250 | |||
XM_011518168.1 | XP_011516470.1 | 9 | 1409 | |||
XM_011518170.1 | XP_011516472.1 | 9 | 1409 | |||
XM_011518172.2 | XP_011516474.1 | 9 | 1409 | |||
XM_017014223.1 | XP_016869712.1 | 9 | 1409 | |||
XM_017014224.1 | XP_016869713.1 | 9 | 1409 | |||
XM_017014225.1 | XP_016869714.1 | 7 | 1095 | |||
XM_017014227.1 | XP_016869716.1 | 8 | 1427 | |||
XM_017014228.1 | XP_016869717.1 | 8 | 1283 | |||
XM_017014229.1 | XP_016869718.1 | 9 | 1409 | |||
XM_017014230.1 | XP_016869719.1 | 9 | 1409 | |||
GenBank mRNA | AF513978.1 | - | 7 | 1106 | ||
AK097636.1 | - | 1 | 14 | |||
AK128098.1 | - | 5 | 651 | |||
AK307387.1 | - | 9 | 1414 | |||
BC089415.1 | - | 9 | 1366 | |||
BC137284.1 | - | 7 | 1130 | 9021281 | ||
BC137286.1 | - | 7 | 1129 | 9021283 |

More Information
Other Species Targeted

Gene Ontology Categories:
