siRNA Details
Gene Symbol: TRIM31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB303153.1 | - | 1 | 197 | ||
AF230386.1 | - | 1 | 306 | |||
AF230387.1 | - | 1 | 306 | |||
Y07828.1 | - | 1 | 306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
