siRNA Details
Gene Symbol: ETNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018638.4 | NP_061108.2 | 4, 5 | 997 | ||
XM_017019580.1 | XP_016875069.1 | 4, 5 | 994 | |||
GenBank mRNA | AF207600.2 | - | 4, 5 | 1063 | ||
AK074425.1 | - | 4, 5 | 942 | |||
AL137750.1 | - | 4, 5 | 946 | |||
BC066907.1 | - | 4, 5 | 997 | 5260109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
