siRNA Details
Gene Symbol: ERLIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007175.6 | NP_009106.1 | 7, 8 | 622 | ||
XM_005273392.2 | XP_005273449.1 | 7, 8 | 782 | |||
XM_006716280.2 | XP_006716343.1 | 6, 7 | 527 | |||
GenBank mRNA | AK291394.1 | - | 7, 8 | 575 | ||
AL442077.1 | - | 609 | ||||
AY358108.1 | - | 7, 8 | 566 | |||
BC005950.1 | - | 7, 8 | 545 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
