siRNA Details
Gene Symbol: ERLIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007175.6 | NP_009106.1 | 12 | 1701 | ||
XM_005273392.2 | XP_005273449.1 | 12 | 1861 | |||
XM_006716280.2 | XP_006716343.1 | 11 | 1606 | |||
GenBank mRNA | AL442077.1 | - | 1688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
